Imidazol-4-carbonsäureethylester 98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Ethyl imidazole-4-carboxylate
H64095.06
H64095.14
H64095.06EA
117
EUR
H64095.06
H64095.14
Imidazol-4-carbonsäureethylester 98%
Imidazol-4-carbonsäureethylester
Purity: 98%
CAS No.: 23785-21-9
Molecular Formula: C6H8N2O2
Formula Weight: 140.14
Storage Temperature: 2°C to 8°C
Physical Form: Solid
Melting Point: 156-160°
Beilstein Reference: 606937
MDL No.: MFCD02646639
Formel:
C₆H₈N₂O₂ Molekulargewicht: 140.14 g/mol Schmelzpunkt: 156…160 °C Lagerungstemperatur: Kühlschrank |
MDL Number:
MFCD07363812 CAS-Nummer: 23785-21-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...