2-Brom-5-chlor-3-nitropyridin 98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
H64065.06
H64065.14
H64065.06EA
92.6
EUR
H64065.06
H64065.14
2-Brom-5-chlor-3-nitropyridin 98%
2-Bromo-5-chloro-3-nitropyridine
Purity: 98%
CAS No.: 75806-86-9
Molecular Formula: C5H2BrClN2O2
Formula Weight: 237.44
Physical Form: Solid
Appearance: Off-white
Melting Point: 175-179°
Beilstein Reference: 147540
UN No.: UN2811
MDL No.: MFCD07368982
Formel:
C₅H₂BrClN₂O₂ Schmelzpunkt: 175…179 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD07368982 CAS-Nummer: 75806-86-9 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...