3-Brom-5-iodpyridin 95%

Lieferant: Thermo Fisher Scientific
Achtung

H64007.03 H64007.06
H64007.03EA 177 EUR
H64007.03 H64007.06
3-Brom-5-iodpyridin 95%
3-Bromo-5-iodopyridine

Purity: 95% CAS No.: 233770-01-9 Molecular Formula: C5H3BrIN Formula Weight: 283.89 Storage Temperature: -30°C to -10°C Physical Form: Solid Sensitivity: Light Sensitive Appearance: Off-white Melting Point: 127-131° Beilstein Reference: 108478 MDL No.: MFCD03086019

Formel: C₅H₃BrIN
Molekulargewicht: 283.89 g/mol
Schmelzpunkt: 127…131 °C
MDL Number: MFCD03086019
CAS-Nummer: 233770-01-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR