Fmoc-3-Cyclohexyl-L-Alanin 98%
Lieferant: Thermo Fisher Scientific
Synonyme:
Fmoc-Cha-OH, N-Fmoc-3-cyclohexyl-L-alanine
H63910.03
H63910.06
H63910.03EA
72.8
EUR
H63910.03
H63910.06
Fmoc-3-Cyclohexyl-L-Alanin 98%
Fmoc-3-Cyclohexyl-L-Alanin
Purity: 98%
CAS No.: 135673-97-1
Molecular Formula: C24H27NO4
Formula Weight: 393.48
Storage Temperature: 2°C to 8°C
Physical Form: Solid
Beilstein Reference: 7052264
MDL No.: MFCD00065614
Formel:
C₂₄H₂₇NO₄ Lagerungstemperatur: Kühlschrank |
MDL Number:
MFCD00065614 CAS-Nummer: 135673-97-1 |