3-Boc-aminoethylazetidine 95%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
3-[2-(Boc-amino)ethyl]azetidine, 3-(N-BOC)-(2-aminoethyl)azetidine, 3-Azetidinecarboxylic acid, 1-(2-aminoethyl)-, 1,1-dimethylethyl ester, 3-Boc-Aminoethylazetidine, 2-Methyl-2-propanyl 1-(2-aminoethyl)-3-azetidinecarboxylate
Purity: 95%
CAS No.: 162696-31-3
Molecular Formula: C10H20N2O2
Formula Weight: 200.28
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: White to pale brown
Melting Point: 84-86°
MDL No.: MFCD09878605
Formel:
C₁₀H₂₀N₂O₂ Schmelzpunkt: 84…86 °C |
MDL Number:
MFCD09878605 CAS-Nummer: 162696-31-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...