(S)-4-Benzyloxycarbonylamino-2-(Fmoc-amino)butyric acid 95%
Lieferant: Thermo Fisher Scientific
Synonyme:
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-4-(((benzyloxy)methyl)amino)butanoic acid, Fmoc-Dab(Z)-OH
(S)-4-Benzyloxycarbonylamino-2-(Fmoc-amino)butyric acid a substituted fluorene compound for proteomics research. Also used in agrochemical, pharmaceutical and dyestuff field.
Purity: 95%
CAS No.: 252049-08-4
Molecular Formula: C27H26N2O6
Formula Weight: 474.51
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Appearance: White
MDL No.: MFCD02259487
Formel:
C₂₇H₂₈N₂O₅ Molekulargewicht: 460.53 g/mol |
MDL Number:
MFCD02259487 CAS-Nummer: 252049-08-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...