Fmoc-3-(1-naphthyl)-D-alanine 98%

Lieferant: Thermo Fisher Scientific

Synonyme: Fmoc-D-1-Nal-OH, N-Fmoc-3-(1-naphthyl)-D-alanine

H63688.03 H63688.06
H63688.03EA 55.5 EUR
H63688.03 H63688.06
Fmoc-3-(1-naphthyl)-D-alanine 98%
Fmoc-3-(1-naphthyl)-D-alanine

Purity: 98% CAS No.: 138774-93-3 Molecular Formula: C28H23NO4 Formula Weight: 437.49 Storage Temperature: 2°C to 8°C Physical Form: Solid Beilstein Reference: 9160026 MDL No.: MFCD00191200

Formel: C₂₈H₂₃NO₄
MDL Number: MFCD00191200
CAS-Nummer: 138774-93-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR