(R)-Methyl-2-amino-3-hydroxypropanoate hydrochloride 95%
Lieferant: Thermo Fisher Scientific
Synonyme:
H-D-Ser-OMe·HCl, H-D-Ser-OMe.HCl, D-Serine methyl ester hydrochloride
H63685.06
H63685.14
H63685.22
H63685.06EA
42.7
EUR
H63685.06
H63685.14
H63685.22
(R)-Methyl-2-amino-3-hydroxypropanoate hydrochloride 95%
(R)-Methyl-2-amino-3-hydroxypropanoate hydrochloride
Purity: 95%
CAS No.: 5874-57-7
Molecular Formula: C4H9NO3.HCl
Formula Weight: 155.58
Physical Form: Solid
Melting Point: 163-166° dec
Optical Rotation: -4°(c=4 in ethanol)
MDL No.: MFCD00066121
Formel:
C₄H₉O₃N·HCl Schmelzpunkt: 163…166 °C (dec.) Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00066121 CAS-Nummer: 5874-57-7 |