Fmoc-N-Me-Val-OH 95%
Lieferant: Thermo Fisher Scientific
Synonyme:
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-N-methyl-L-valine, Fmoc-N-Me-Val-OH, N-Fmoc-N-methyl-L-valine
H63671.03
H63671.06
H63671.03EA
106
EUR
H63671.03
H63671.06
Fmoc-N-Me-Val-OH 95%
Fmoc-N-Me-Val-OH
Purity: 95%
CAS No.: 84000-11-3
Molecular Formula: C21H23NO4
Formula Weight: 353.41
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: White
Melting Point: 187-190°
Optical Rotation: -68°(c=1 in DMF)
MDL No.: MFCD00153395
Formel:
C₂₁H₂₃NO₄ Molekulargewicht: 353.418 g/mol Schmelzpunkt: 187…190 °C |
MDL Number:
MFCD00153395 CAS-Nummer: 84000-11-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...