8-[(9H-Fluoren-9-ylmethoxy)carbonylamino]-3,6-dioxa-n-octanoic acid 95%
Lieferant: Thermo Fisher Scientific
Synonyme:
Fmoc-NH-PEG2-CH2COOH, 8-(Fmoc-amino)-3,6-dioxa-n-octanoic acid, Fmoc-8-amino-3,6-dioxaoctanoic acid, [2-[2-[(Fmoc-amino)ethoxy]ethoxy]acetic acid, 1-(9H-Fluoren-9-yl)-3-oxo-2,7,10-trioxa-4-azadodecan-12-oic acid, [2-[2-[(9H-Fluoren-9-ylmethoxy)carbonylamino]ethoxy]ethoxy]acetic acid
Purity: 95%
CAS No.: 166108-71-0
Molecular Formula: C21H23NO6
Formula Weight: 385.42
Physical Form: Crystalline powder
Appearance: White
Beilstein Reference: 9658732
MDL No.: MFCD01321015
Formel:
C₂₁H₂₃NO₆ Molekulargewicht: 385.42 g/mol Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD01321015 CAS-Nummer: 166108-71-0 |