Fmoc-Lys(biotin)-OH 95%

Lieferant: Thermo Fisher Scientific

Synonyme: Nα-Biotinyl-Nepsilon-Fmoc-L-lysine, Fmoc-Biocytin, (S)-2-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)-6-(5-((3aS,4S,6aR)-2-oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)pentanamido)hexanoic acid, Nalpha-[(9H-Fluoren-9-ylmethoxy)carbonyl]-Nepsilon-biotinyl-L-lysine, Fmoc-Lys(biotinyl)-OH, Nalpha-Fmoc-Nepsilon-biotinyl-L-lysine

H63609.03 H63609.06
H63609.03EA 350 EUR
H63609.03 H63609.06
Fmoc-Lys(biotin)-OH 95%
Fmoc-Lys(biotin)-OH

Purity: 95% CAS No.: 146987-10-2 Molecular Formula: C31H38N4O6S Formula Weight: 594.72 Physical Form: Solid MDL No.: MFCD00270741

Formel: C₃₁H₃₈N₄O₆S
Molekulargewicht: 594.727 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00270741
CAS-Nummer: 146987-10-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR