4-Chlor-L-phenylalanin 97%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
L-4-Chlorophenylalanine, H-Phe(4-Cl)-OH
H63573.MD
H63573.03
H63573.06
H63573.MDEA
75.6
EUR
H63573.MD
H63573.03
H63573.06
4-Chlor-L-phenylalanin 97%
4-Chlor-L-phenylalanin
Purity: 97%
CAS No.: 14173-39-8
Molecular Formula: C9H10ClNO2
Formula Weight: 199.63
Storage Temperature: -30°C to -10°C
Physical Form: Solid
Melting Point: 235-238°
Beilstein Reference: 2416150
UN No.: UN2811
MDL No.: MFCD00063065
Formel:
C₉H₁₀ClNO₂ Schmelzpunkt: 235…238 °C |
MDL Number:
MFCD00063065 CAS-Nummer: 14173-39-8 EINECS: 238-023-5 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...