N-Acetyl-D-alanine 98%
Lieferant: Thermo Fisher Scientific
Synonyme:
N-Ac-D-Ala-OH, Ac-D-Ala-OH
H63450.03
H63450.06
H63450.03EA
47.8
EUR
H63450.03
H63450.06
N-Acetyl-D-alanine 98%
N-Acetyl-D-alanine
Purity: 98%
CAS No.: 19436-52-3
Molecular Formula: C5H9NO3
Formula Weight: 131.13
Storage Temperature: -30°C to -10°C
Physical Form: Solid
MDL No.: MFCD00063133
Formel:
C₅H₉NO₃ |
MDL Number:
MFCD00063133 CAS-Nummer: 19436-52-3 EINECS: 243-066-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...