L-Alanine-4-nitroanilide hydrochloride 98%
Lieferant: Thermo Fisher Scientific
Synonyme:
H-Ala-pNA·HCl, L-Alanine 4-nitroanilide hydrochloride
H63393.MD
H63393.03
H63393.06
H63393.MDEA
53.7
EUR
H63393.MD
H63393.03
H63393.06
L-Alanine-4-nitroanilide hydrochloride 98%
L-Alanine-4-nitroanilide hydrochloride
Purity: 98%
CAS No.: 31796-55-1
Molecular Formula: C9H11N3O3.HCl
Formula Weight: 245.66
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Appearance: Pale yellow
Beilstein Reference: 6303731
MDL No.: MFCD00039088
Formel:
C₉H₁₂ClN₃O₃ |
MDL Number:
MFCD00039088 CAS-Nummer: 31796-55-1 EINECS: 250-811-0 |