N-Fmoc-4-iodo-D-phenylalanine 98%
Lieferant: Thermo Fisher Scientific
Synonyme:
Fmoc-D-Phe(4-I)-OH
H63292.03
H63292.06
H63292.03EA
79.7
EUR
H63292.03
H63292.06
N-Fmoc-4-iodo-D-phenylalanine 98%
N-Fmoc-4-iodo-D-phenylalanine
Purity: 98%
CAS No.: 205526-29-0
Molecular Formula: C24H20INO4
Formula Weight: 513.32
Storage Temperature: 2°C to 8°C
Physical Form: Solid
Sensitivity: Light Sensitive
Beilstein Reference: 9159551
MDL No.: MFCD00672557
Formel:
C₂₄H₂₀INO₄ |
MDL Number:
MFCD00672557 CAS-Nummer: 205526-29-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...