H-Asp(OtBu)-OMe·HCl 98%

Lieferant: Thermo Fisher Scientific

Synonyme: L-Aspartic acid 4-tert-butyl 1-methyl ester hydrochloride, H-Asp(OtBu)-OMe.HCl

H63257.03 H63257.06
H63257.03EA 88.6 EUR
H63257.03 H63257.06
H-Asp(OtBu)-OMe·HCl 98%
H-Asp(OtBu)-OMe·HCl

Purity: 98% CAS No.: 2673-19-0 Molecular Formula: C9H17NO4.HCl Formula Weight: 239.7 Storage Temperature: 2°C to 8°C Physical Form: Solid Beilstein Reference: 6005591 MDL No.: MFCD00153427

Formel: C₉H₁₈ClNO₄
MDL Number: MFCD00153427
CAS-Nummer: 2673-19-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR