5-bromoquinolin-6-amine 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
H63150.03
H63150.06
H63150.03EA
193
EUR
H63150.03
H63150.06
5-bromoquinolin-6-amine 97%
5-bromoquinolin-6-amine
Purity: 97%
CAS No.: 50358-42-4
Molecular Formula: C9H7BrN2
Formula Weight: 223.07
Physical Form: Solid
Appearance: Yellow to brown
MDL No.: MFCD00234503
Formel:
C₉H₇BrN₂ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00234503 CAS-Nummer: 50358-42-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...