Fmoc-Lys(Dnp)-OH 98%
Lieferant: Thermo Fisher Scientific
Synonyme:
Nepsilon-2,4-Dinitrophenyl-Nα-Fmoc-L-lysine
H63050.03
H63050.06
H63050.03EA
173
EUR
H63050.03
H63050.06
Fmoc-Lys(Dnp)-OH 98%
Fmoc-Lys(Dnp)-OH
Purity: 98%
CAS No.: 148083-64-1
Molecular Formula: C27H26N4O8
Formula Weight: 534.52
Physical Form: Solid
MDL No.: MFCD00273454
Formel:
C₂₇H₂₆N₄O₈ Molekulargewicht: 534.525 g/mol Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00273454 CAS-Nummer: 148083-64-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...