3-(1-Methyl-4-piperidinyl)indole 97%

Lieferant: Thermo Fisher Scientific
Achtung

H63020.MD H63020.06
H63020.MDEA 106 EUR
H63020.MD H63020.06
3-(1-Methyl-4-piperidinyl)indole 97%
3-(1-Methyl-4-piperidinyl)indole

Purity: 97% CAS No.: 17403-07-5 Molecular Formula: C14H18N2 Formula Weight: 214.31 Physical Form: Solid Appearance: Off-white to yellow Beilstein Reference: 1573327 MDL No.: MFCD07776765

Formel: C₁₄H₁₈N₂
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD07776765
CAS-Nummer: 17403-07-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR