Boc-N-Me-Val-OH 95%
Lieferant: Thermo Fisher Scientific
Synonyme:
N-(tert-Butoxycarbonyl)-N-methyl-L-valine, N-Boc-N-methyl-L-valine, Boc-N-Me-Val-OH
H62771.03
H62771.06
H62771.03EA
86.5
EUR
H62771.03
H62771.06
Boc-N-Me-Val-OH 95%
Boc-N-Me-Val-OH
Purity: 95%
CAS No.: 45170-31-8
Molecular Formula: C11H21NO4
Formula Weight: 231.29
Physical Form: Solid
Beilstein Reference: 2646180
Optical Rotation: -94°(c=0.5 in ethanol)
MDL No.: MFCD00038760
Formel:
C₁₁H₂₁NO₄ Molekulargewicht: 231.292 g/mol Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00038760 CAS-Nummer: 45170-31-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...