Boc-N-Me-D-Phe-OH 98%
Lieferant: Thermo Fisher Scientific
Synonyme:
N-Boc-N-methyl-D-phenylalanine, N-(tert-Butoxycarbonyl)-N-methyl-D-phenylalanine, Boc-N-Me-D-Phe-OH
H62638.03
H62638.06
H62638.03EA
139
EUR
H62638.03
H62638.06
Boc-N-Me-D-Phe-OH 98%
Boc-N-Me-D-Phe-OH
Purity: 98%
CAS No.: 85466-66-6
Molecular Formula: C15H21NO4
Formula Weight: 279.33
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: White
MDL No.: MFCD00151890
Formel:
C₁₅H₂₁NO₄ |
MDL Number:
MFCD00151890 CAS-Nummer: 85466-66-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...