Fmoc-D-Dap(Boc)-OH 95%

Lieferant: Thermo Fisher Scientific

Synonyme: (R)-3-(Boc-amino)-2-(Fmoc-amino)propionic acid

H62381.03 H62381.06
H62381.03EA 190 EUR
H62381.03 H62381.06
Fmoc-D-Dap(Boc)-OH 95%
Fmoc-D-Dap(Boc)-OH

Purity: 95% CAS No.: 198544-42-2 Molecular Formula: C23H26N2O6 Formula Weight: 426.46 Physical Form: Solid MDL No.: MFCD00798648

Formel: C₂₃H₂₆N₂O₆
Molekulargewicht: 426.469 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00798648
CAS-Nummer: 198544-42-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR