Dimethyl-N-(tert-butoxycarbonyl)-L-glutamate 97%

Lieferant: Thermo Fisher Scientific

Synonyme: N-Boc-L-glutamic Acid Dimethyl Ester, N-Boc-L-glutamic acid 1,5-dimethyl ester, Dimethyl N-Boc-L-glutamate, (S)-Dimethyl 2-((tert-butoxycarbonyl)amino)pentanedioate, N-(tert-Butoxycarbonyl)-L-glutamic Acid Dimethyl Ester, Boc-Glu(OMe)-OMe

H62056.06 H62056.14
H62056.06EA 88.3 EUR
H62056.06 H62056.14
Dimethyl-N-(tert-butoxycarbonyl)-L-glutamate 97%
Dimethyl-N-(tert-butoxycarbonyl)-L-glutamate

Purity: 97% CAS No.: 59279-60-6 Molecular Formula: C12H21NO6 Formula Weight: 275.30 Physical Form: Liquid Appearance: Colourless to pale yellow MDL No.: MFCD03094774

Formel: C₁₂H₂₁NO₆
Molekulargewicht: 275.30 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD03094774
CAS-Nummer: 59279-60-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR