3-Benzo[b]thiophensulfonylchlorid 97%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
1-Benzothiophene-3-sulphonyl Chloride, Benzothiophene-3-sulphonyl chloride
H61368.MD
H61368.03
H61368.MDEA
85.6
EUR
H61368.MD
H61368.03
3-Benzo[b]thiophensulfonylchlorid 97%
3-Benzo[b]thiophensulfonylchlorid
Purity: 97%
CAS No.: 18494-87-6
Molecular Formula: C8H5ClO2S2
Formula Weight: 232.71
Storage Temperature: 2°C to 8°C
Melting Point: 86-88°
Beilstein Reference: 1285265
UN No.: UN3261
MDL No.: MFCD02681897
Formel:
C₈H₅ClO₂S₂ Schmelzpunkt: 86…88 °C |
MDL Number:
MFCD02681897 CAS-Nummer: 18494-87-6 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...