6-(Boc-amino)-capronsäure 95%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
H61350.06
H61350.14
H61350.06EA
97.1
EUR
H61350.06
H61350.14
6-(Boc-amino)-capronsäure 95%
6-(Boc-amino)-capronsäure
Purity: 95%
CAS No.: 6404-29-1
Molecular Formula: C11H21NO4
Formula Weight: 231.29
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: White
Melting Point: 35-40°
Beilstein Reference: 2049561
MDL No.: MFCD00037798
Formel:
C₁₁H₂₁NO₄ Schmelzpunkt: 35…40 °C Lagerungstemperatur: Kühlschrank |
MDL Number:
MFCD00037798 CAS-Nummer: 6404-29-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...