Boc-D-Pyr-OH 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
(R)-1-(tert-Butoxycarbonyl)-5-oxopyrrolidine-2-carboxylic acid
H58701.03
H58701.06
H58701.03EA
49.7
EUR
H58701.03
H58701.06
Boc-D-Pyr-OH 97%
Boc-D-Pyr-OH
Purity: 97%
CAS No.: 160347-90-0
Molecular Formula: C10H15NO5
Formula Weight: 229.23
Storage Temperature: 2°C to 8°C
Melting Point: 111-116°
Optical Rotation: +35° (c=1 in chloroform)
MDL No.: MFCD08458561
Formel:
C₁₀H₁₅NO₅ Molekulargewicht: 229.232 g/mol Schmelzpunkt: 111…116 °C |
MDL Number:
MFCD08458561 CAS-Nummer: 160347-90-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...