2-Oxa-7-azaspiro[3.5]nonane hemioxalate 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
H57834.MD
H57834.ME
H57834.MDEA
325
EUR
H57834.MD
H57834.ME
2-Oxa-7-azaspiro[3.5]nonane hemioxalate 97%
2-Oxa-7-azaspiro[3.5]nonane hemioxalate
Purity: 97%
CAS No.: 1429056-28-9
Molecular Formula: C7H13NO.0.5C2H2O4
Formula Weight: 172.20
Storage Temperature: 2°C to 8°C
Physical Form: Solid
Density: 1.048
Appearance: White to grey
Melting Point: 208°
MDL No.: MFCD11035888
Formel:
C₇H₁₃NO·½C₂H₂O₄ Schmelzpunkt: 208 °C Dichte: 1,048 |
MDL Number:
MFCD11035888 CAS-Nummer: 1429056-28-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...