N-Boc-β-phenyl-D-phenylalanine 98%
Lieferant: Thermo Fisher Scientific
Synonyme:
N-(tert-Butoxycarbonyl)-beta-phenyl-D-phenylalanine, Boc-D-Ala(3,3-diphenyl)-OH, Boc-D-3,3-diphenylalanine
H56885.ME
H56885.03
H56885.MEEA
166
EUR
H56885.ME
H56885.03
N-Boc-β-phenyl-D-phenylalanine 98%
N-Boc-beta-phenyl-D-phenylalanine
Purity: 98%
CAS No.: 143060-31-5
Molecular Formula: C20H23NO4
Formula Weight: 341.40
Melting Point: 157°
Optical Rotation: -30 (c=1 in ethanol)
MDL No.: MFCD00191187
Formel:
C₂₀H₂₃NO₄ Schmelzpunkt: 157 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00191187 CAS-Nummer: 143060-31-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...