Methyl-1H-imidazol-5-carboxylat 98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Methyl 4-imidazolecarboxylate, Methyl 1H-imidazole-5-carboxylate

H55932.03 H55932.06
H55932.03EA 123 EUR
H55932.03 H55932.06
Methyl-1H-imidazol-5-carboxylat 98%
Methyl-1H-imidazole-5-carboxylate

Purity: 98% CAS No.: 17325-26-7 Molecular Formula: C5H6N2O2 Formula Weight: 126.11 Melting Point: 154-156° MDL No.: MFCD00216584

Formel: C₅H₆N₂O₂
Molekulargewicht: 126.11 g/mol
Schmelzpunkt: 154…156 °C
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00216584
CAS-Nummer: 17325-26-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR