Triphenyl borate 97%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
Phenyl Borate, Boric Acid Triphenyl Ester
H55686.06
H55686.14
H55686.06EA
33.8
EUR
H55686.06
H55686.14
Triphenyl borate 97%
Triphenyl borate
Purity: 97%
CAS No.: 1095-03-0
Molecular Formula: C18H15BO3
Formula Weight: 290.13
Physical Form: Solid
Sensitivity: Moisture Sensitive
Melting Point: 98-101°
UN No.: UN2811
MDL No.: MFCD00059011
Formel:
C₁₈H₁₅BO₃ Schmelzpunkt: 98…101 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00059011 CAS-Nummer: 1095-03-0 EINECS: 214-137-0 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...