1-Benzyl-N-carbobenzoxy-D-glutamate 97%
Lieferant: Thermo Fisher Scientific
Synonyme:
1-Benzyl N-Cbz-D-glutamate, Z-D-Glu-OBzl, N-CBZ-D-glutamic acid alpha-benzyl ester, N-Carbobenzoxy-D-glutamic Acid 1-Benzyl Ester, N-Cbz-D-glutamic Acid 1-Benzyl Ester, Z-D-Glu-Obzl
H55121.03
H55121.06
H55121.03EA
61
EUR
H55121.03
H55121.06
1-Benzyl-N-carbobenzoxy-D-glutamate 97%
1-Benzyl-N-carbobenzoxy-D-glutamate
Purity: 97%
CAS No.: 65706-99-2
Molecular Formula: C20H21NO6
Formula Weight: 371.39
Beilstein Reference: 5305848
MDL No.: MFCD00069647
Formel:
C₂₀H₂₁NO₆ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00069647 CAS-Nummer: 65706-99-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...