tert-Butyl-3-hydroxyphenylcarbamate 97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: tert-Butyl (3-hydroxyphenyl)carbamate

H55059.14 H55059.06
H55059.14EA 265 EUR
H55059.14 H55059.06
tert-Butyl-3-hydroxyphenylcarbamate 97%
tert-Butyl-3-hydroxyphenylcarbamate

Purity: 97% CAS No.: 19962-06-2 Molecular Formula: C11H15NO3 Formula Weight: 209.25 Melting Point: 134-138° MDL No.: MFCD06798070

Formel: C₁₁H₁₅NO₃
Schmelzpunkt: 134…138 °C
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD06798070
CAS-Nummer: 19962-06-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR