4-[(2,4-Dimethoxyphenyl)(Fmoc-amino)methyl]phenoxyacetic acid ≥98%
Lieferant: Thermo Fisher Scientific
Synonyme:
Rink amide linker, FMOC-AM, Fmoc-2,4-dimethoxy-4'-(carboxymethyloxy)-benzhydrylamine, 2-(4-(((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)(2,4-dimethoxyphenyl)methyl)phenoxy)acetic acid, 4-[(2,4-Dimethoxyphenyl)[(9H-fluoren-9-ylmethoxy)carbonylamino]methyl]phenoxyacetic acid
Purity: 98+%
CAS No.: 145069-56-3
Molecular Formula: C32H29NO7
Formula Weight: 539.58
Storage Temperature: 2°C to 8°C
Melting Point: ca 180° dec.
MDL No.: MFCD00153509
Formel:
C₃₂H₂₉NO₇ Molekulargewicht: 539.58 g/mol Schmelzpunkt: ∼ 180 °C (Zers.) |
MDL Number:
MFCD00153509 CAS-Nummer: 126828-35-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...