4-(6-Amino-3-pyridyl)-1-boc-piperazine 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
1-Boc-4-(6-amino-3-pyridyl)piperazine, tert-Butyl 4-(6-Amino-3-pyridyl)piperazine-1-carboxylate, tert-Butyl 4-(6-aminopyridin-3-yl)piperazine-1-carboxylate, 1-tert-Butoxycarbonyl-4-(6-amino-3-pyridyl)piperazine, 4-(6-Amino-3-pyridyl)piperazine-1-carboxylic acid tert-Butyl ester
Purity: 97%
CAS No.: 571188-59-5
Molecular Formula: C14H22N2O4
Formula Weight: 278.35
Storage Temperature: 2°C to 8°C
Beilstein Reference: 10173494
MDL No.: MFCD11594962
Formel:
C₁₄H₂₂N₂O₄ |
MDL Number:
MFCD11594962 CAS-Nummer: 571188-59-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...