[(tert-Butoxycarbonyl)aminooxy]acetic acid ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: N-Boc-(carboxymethoxy)amine, Boc-Aoa-OH, (Boc-aminooxy)acetic Acid

H54109.03 H54109.06 H54109.14
H54109.03EA 69.5 EUR
H54109.03 H54109.06 H54109.14
[(tert-Butoxycarbonyl)aminooxy]acetic acid ≥98%
[(tert-Butoxycarbonyl)aminooxy]acetic acid

Purity: 98+% CAS No.: 42989-85-5 Molecular Formula: C7H13NO5 Formula Weight: 191.18 Melting Point: ca 115° dec. Beilstein Reference: 6137646 MDL No.: MFCD01632027

Formel: C₇H₁₃NO₅
Molekulargewicht: 191.18 g/mol
Schmelzpunkt: ∼ 115 °C (Zers.)
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD01632027
CAS-Nummer: 42989-85-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR