N,N'-Bis-Z-1-guanylpyrazole ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
H54092.03
H54092.06
H54092.03EA
117
EUR
H54092.03
H54092.06
N,N'-Bis-Z-1-guanylpyrazole ≥98%
N,N'-Bis-Z-1-guanylpyrazole
Purity: 98+%
CAS No.: 152120-55-3
Molecular Formula: C20H18N4O4
Formula Weight: 378.38
Melting Point: 87-91°
MDL No.: MFCD02683516
Formel:
C₂₀H₁₈N₄O₄ Schmelzpunkt: 87…91 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD02683516 CAS-Nummer: 152120-55-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...