Boc-aza-L-leucine 97%

Lieferant: Thermo Fisher Scientific

Synonyme: N-(tert-Butoxycarbonyl)-3-(dimethylamino)-L-alanine

H52810.MD H52810.03
H52810.MDEA 144 EUR
H52810.MD H52810.03
Boc-aza-L-leucine 97%
Boc-aza-L-leucine

Purity: 97% CAS No.: 94778-71-9 Molecular Formula: C10H20N2O4 Formula Weight: 232.28 MDL No.: MFCD02682423

Formel: C₁₀H₂₀N₂O₄
Molekulargewicht: 232.28 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD02682423
CAS-Nummer: 94778-71-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR