2-Carbamoylbenzeneboronic acid 96%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
2-Aminocarbonylphenylboronic acid
H52642.MD
H52642.03
H52642.MDEA
111
EUR
H52642.MD
H52642.03
2-Carbamoylbenzeneboronic acid 96%
2-Carbamoylbenzeneboronic acid
Purity: 96%
CAS No.: 380430-54-6
Molecular Formula: C7H8BNO3
Formula Weight: 164.96
Melting Point: 226-230°
MDL No.: MFCD02179454
Formel:
C₇H₈BNO₃ Schmelzpunkt: 226…230 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD02179454 CAS-Nummer: 380430-54-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...