Boc-L-β-homoaspartic acid-γ-benzyl ester 95%
Lieferant: Thermo Fisher Scientific
Synonyme:
Boc-β-HomoAsp(OBzl)-OH, Boc-β-HoAsp(OBzl)-OH
H52191.MD
H52191.03
H52191.MDEA
158
EUR
H52191.MD
H52191.03
Boc-L-β-homoaspartic acid-γ-benzyl ester 95%
Boc-L-beta-homoaspartic acid-gamma-benzyl ester
Purity: 95%
CAS No.: 254101-10-5
Molecular Formula: C17H23NO6
Formula Weight: 337.37
Beilstein Reference: 8430789
MDL No.: MFCD01862861
Formel:
C₁₇H₂₃NO₆ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD01862861 CAS-Nummer: 254101-10-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...