Fmoc-L-propargylglycine 95%
Lieferant: Thermo Fisher Scientific
Synonyme:
Fmoc-Pra-OH, N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-propargylglycine, N-Fmoc-L-propargylglycine, (S)-2-(Fmoc-amino)-4-pentynoic acid, Fmoc-propargyl-Gly-OH
H52171.MD
H52171.03
H52171.06
H52171.MDEA
118
EUR
H52171.MD
H52171.03
H52171.06
Fmoc-L-propargylglycine 95%
Fmoc-L-propargylglycine
Purity: 95%
CAS No.: 198561-07-8
Molecular Formula: C20H17NO4
Formula Weight: 335.36
Beilstein Reference: 9151699
MDL No.: MFCD01075095
Formel:
C₂₀H₁₇NO₄ Molekulargewicht: 335.359 g/mol Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD01075095 CAS-Nummer: 198561-07-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...