Nα-Fmoc-Nω-(2,2,4,6,7-pentamethyl-2,3-dihydrobenzo[b]furan-5-ylsulphonyl)-L-β-homoarginine 95%

Lieferant: Thermo Fisher Scientific

Synonyme: Fmoc-β-HomoArg(Pbf)-OH

H52115.MD H52115.03
H52115.MDEA 110 EUR
H52115.MD H52115.03
Nα-Fmoc-Nω-(2,2,4,6,7-pentamethyl-2,3-dihydrobenzo[b]furan-5-ylsulphonyl)-L-β-homoarginine 95%
Nα-Fmoc-Nω-(2,2,4,6,7-pentamethyl-2,3-dihydrobenzo[b]furan-5-ylsulphonyl)-L-β-homoarginine

Purity: 95% CAS No.: 401915-53-5 Molecular Formula: C35H42N4O7S Formula Weight: 662.80 MDL No.: MFCD03788172

Formel: C₃₅H₄₂N₄O₇S
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD03788172
CAS-Nummer: 401915-53-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR