Fmoc-D-4,4'-biphenylalanine 95%
Lieferant: Thermo Fisher Scientific
Synonyme:
Fmoc-4-phenyl-D-Phe-OH, Fmoc-D-Bip(4,4’)-OH, Fmoc-D-Bip(4,4')-OH
H52108.03
H52108.06
H52108.03EA
172
EUR
H52108.03
H52108.06
Fmoc-D-4,4'-biphenylalanine 95%
Fmoc-D-4,4'-biphenylalanine
Purity: 95%
CAS No.: 205526-38-1
Molecular Formula: C30H25NO4
Formula Weight: 463.52
MDL No.: MFCD00191199
Formel:
C₃₀H₂₅NO₄ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00191199 CAS-Nummer: 205526-38-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...