(S)-3-(4-Aminophenyl)-2-((tert-butoxycarbonyl)amino)propanoic acid 95%

Lieferant: Thermo Fisher Scientific

H51980.06 H51980.03
H51980.06EA 468 EUR
H51980.06 H51980.03
(S)-3-(4-Aminophenyl)-2-((tert-butoxycarbonyl)amino)propanoic acid 95%
(S)-3-(4-Aminophenyl)-2-((tert-butoxycarbonyl)amino)propanoic acid

Purity: 95% CAS No.: 55533-24-9 Molecular Formula: C14H20N2O4 Formula Weight: 280.32 Melting Point: 124-126° MDL No.: MFCD00038139

Formel: C₁₄H₂₀N₂O₄
Schmelzpunkt: 124…126 °C
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00038139
CAS-Nummer: 55533-24-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR