Fmoc-L-β-homoleucine 95%

Lieferant: Thermo Fisher Scientific

Synonyme: Fmoc-L-β-HoLeu-OH, (S)-3-[[(9H-Fluoren-9-ylmethoxy)carbonyl]amino]-5-methylhexanoic acid, Fmoc-L-beta-homoleucine, Fmoc-β-HomoLeu-OH, Fmoc-beta-Homoleu-OH, [(9H-Fluoren-9-ylmethoxy)carbonyl]-L-beta-homoleucine

H51976.MD H51976.03
H51976.MDEA 133 EUR
H51976.MD H51976.03
Fmoc-L-β-homoleucine 95%
Fmoc-L-beta-homoleucine

Purity: 95% CAS No.: 193887-44-4 Molecular Formula: C22H25NO4 Formula Weight: 367.44 Beilstein Reference: 7723942 MDL No.: MFCD01863059

Formel: C₂₂H₂₅NO₄
Molekulargewicht: 367.45 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD01863059
CAS-Nummer: 193887-44-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR