Fmoc-3-(2-naphthyl)-D-alanine 95%
Lieferant: Thermo Fisher Scientific
Synonyme:
Fmoc-D-2-Nal-OH
H51974.03
H51974.06
H51974.03EA
155
EUR
H51974.03
H51974.06
Fmoc-3-(2-naphthyl)-D-alanine 95%
Fmoc-3-(2-naphthyl)-D-alanine
Purity: 95%
CAS No.: 138774-94-4
Molecular Formula: C28H23NO4
Formula Weight: 437.49
Beilstein Reference: 7864489
MDL No.: MFCD00077044
Formel:
C₂₈H₂₃NO₄ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00077044 CAS-Nummer: 138774-94-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...