(S)-2-((tert-Butoxycarbonyl)amino)-3-(4-iodophenyl)propanoic acid 98%

Lieferant: Thermo Fisher Scientific

Synonyme: Boc-Phe(4-I)-OH, Boc-p-iodo-Phe-OH

H51960.06 H51960.14
H51960.06EA 158 EUR
H51960.06 H51960.14
(S)-2-((tert-Butoxycarbonyl)amino)-3-(4-iodophenyl)propanoic acid 98%
(S)-2-((tert-Butoxycarbonyl)amino)-3-(4-iodophenyl)propanoic acid

Purity: 98% CAS No.: 62129-44-6 Molecular Formula: C14H18INO4 Formula Weight: 391.21 Melting Point: 150° dec. Beilstein Reference: 4503851 Optical Rotation: +22.5° (c=1 in ethyl acetate) MDL No.: MFCD00037119

Formel: C₁₄H₁₈INO₄
Molekulargewicht: 391.2 g/mol
Schmelzpunkt: 150 °C (Zers.)
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00037119
CAS-Nummer: 62129-44-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR