Thiophene-2,5-diboronic acid bis(pinacol) ester 97%

Lieferant: Thermo Fisher Scientific

H51725.03 H51725.06
H51725.03EA 78.2 EUR
H51725.03 H51725.06
Thiophene-2,5-diboronic acid bis(pinacol) ester 97%
Thiophene-2,5-diboronic acid bis(pinacol) ester

Purity: 97% CAS No.: 175361-81-6 Molecular Formula: C16H26B2O4S Formula Weight: 336.06 Melting Point: 216-220° MDL No.: MFCD09800563

Formel: C₁₆H₂₆B₂O₄S
Schmelzpunkt: 216…220 °C
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD09800563
CAS-Nummer: 175361-81-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR