2-Cyclopropylmethoxy-3-iodo-pyridine 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
H50084.03
H50084.06
H50084.03EA
111
EUR
H50084.03
H50084.06
2-Cyclopropylmethoxy-3-iodo-pyridine 97%
2-Cyclopropylmethoxy-3-iodopyridine
Purity: 97%
CAS No.: 766557-62-4
Molecular Formula: C9H10INO
Formula Weight: 275.09
MDL No.: MFCD06659010
Formel:
C₉H₁₀INO Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD06659010 CAS-Nummer: 766557-62-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...