7-Azaindole-5-boronic acid pinacol ester 97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 7-Azaindole-5-boronic Acid Pinacol Ester

H50044.03 H50044.MD
H50044.03EA 136 EUR
H50044.03 H50044.MD
7-Azaindole-5-boronic acid pinacol ester 97%
7-Azaindole-5-boronic acid pinacol ester

Purity: 97% CAS No.: 754214-56-7 Molecular Formula: C13H17BN2O2 Formula Weight: 244.10 Melting Point: 241-245° MDL No.: MFCD08060937

Formel: C₁₃H₁₇BN₂O₂
Schmelzpunkt: 241…245 °C
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD08060937
CAS-Nummer: 754214-56-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR