Ethyl 2-amino-5,6-dihydro-4H-cyclopenta[b]-thiophene-3-carboxylate 96%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 2-Amino-5,6-dihydro-4-[4H]-cyclopenta[B]thiophene-3-carboxylic acid ethyl ester, Ethyl 2-amino-5,6-dihydro-4H-cyclopenta[b]thiophene-3-carboxylate

H33196.06 H33196.14
H33196.06EA 90.9 EUR
H33196.06 H33196.14
Ethyl 2-amino-5,6-dihydro-4H-cyclopenta[b]-thiophene-3-carboxylate 96%
Ethyl-2-amino-5,6-dihydro-4H-cyclopenta[b]thiophene-3-carboxylate

Purity: 96% CAS No.: 4815-29-6 Molecular Formula: C10H13NO2S Formula Weight: 211.28 Melting Point: 95-96° MDL No.: MFCD00102063

Formel: C₁₀H₁₃NO₂S
Schmelzpunkt: 95…96 °C
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00102063
CAS-Nummer: 4815-29-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR